Tisk…

S-Benzyl-L-cysteine 99%

Dodavatel: Thermo Fisher Scientific

Synonyms: H-Cys(Bzl)-OH

L11666.06 L11666.14
L11666.06EA 718 CZK
L11666.06 L11666.14
S-Benzyl-L-cysteine 99%
S-Benzyl-L-cysteine

Purity: 99% CAS No.: 3054-01-1 Molecular Formula: C10H13NO2S Formula Weight: 211.28 Melting Point: ca 214° dec. Beilstein Reference: 1879358 Optical Rotation: +28° (c=1.5 in 1N NaOH) MDL No.: MFCD00002613

Vzorec: C₁₀H₁₃NO₂S
Storage Temperature: Ambient
MDL Number: MFCD00002613
CAS číslo: 3054-01-1
EINECS: 221-273-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR