Tisk…

N-(2-Cyanoethyl)morpholine ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Morpholin-4-ylpropanenitrile, 3-Morpholinopropionitrile, 3-(4-Morpholino)propionitrile, 3-(morpholin-4-yl)propanenitrile

L11664.06 L11664.14
L11664.06EA 900 CZK
L11664.06 L11664.14
N-(2-Cyanoethyl)morpholine ≥98%
N-(2-Cyanoethyl)morpholine

Purity: 98+% CAS No.: 4542-47-6 Molecular Formula: C7H12N2O Formula Weight: 140.19 Flash Point: 110°(230°F) Refractive Index: 1.4720 Melting Point: 20-21° Boiling Point: 106-109°/7mm Beilstein Reference: 110425 UN No.: UN3276 MDL No.: MFCD00006178

Vzorec: C₇H₁₂N₂O
MW: 140.19 g/mol
Bod varu: 106…109 °C (7 mmHg)
Bod tání: 20…21 °C
Hustota: 1.037
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006178
CAS číslo: 4542-47-6
EINECS: 224-894-9
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR