Allyl chloroacetate 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L11531.06
L11531.14
L11531.06EA
851
CZK
L11531.06
L11531.14
Allyl chloroacetate 98%
Allyl chloroacetate
Purity: 98%
CAS No.: 2916-14-5
Molecular Formula: ClCH2CO2CH2CH=CH2
Formula Weight: 134.56
Flash Point: 61°(141°F)
Refractive Index: 1.4450
Boiling Point: 162-164°
Beilstein Reference: 1747000
UN No.: UN2810
MDL No.: MFCD00058936
Vzorec:
C₅H₇ClO₂ Bod varu: 162…164 °C Hustota: 1,160 Bod vzplanutí: 61 °C (141 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00058936 CAS číslo: 2916-14-5 EINECS: 220-839-8 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...