Tisk…

4-(9H-Carbazol-9-yl)aniline hydrochloride 98%

Dodavatel: Thermo Fisher Scientific

L11362.03 L11362.MD
L11362.03EA 1880 CZK
L11362.03 L11362.MD
4-(9H-Carbazol-9-yl)aniline hydrochloride 98%
4-(9H-Carbazol-9-yl)aniline hydrochloride

Purity: 98% CAS No.: 312700-07-5 Molecular Formula: C18H14N2.HCl Formula Weight: 294.79 Melting Point: ca 250° dec. MDL No.: MFCD00082678

Vzorec: C₁₈H₁₅ClN₂
MW: 294.79 g/mol
Bod tání: ∼ 250 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00082678
CAS číslo: 312700-07-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR