N-(2-Hydroxyethyl)succinimide 95%
Dodavatel: Thermo Scientific
|
Varování
|
L11306.06
L11306.14
L11306.06EA
1300
CZK
L11306.06
L11306.14
N-(2-Hydroxyethyl)succinimide 95%
N-(2-Hydroxyethyl)succinimide
Purity: 95%
CAS No.: 18190-44-8
Molecular Formula: C6H9NO3
Formula Weight: 143.14
Melting Point: 59-61°
Beilstein Reference: 124744
MDL No.: MFCD00078332
Vzorec:
C₆H₉NO₃ MW: 143,14 g/mol Bod tání: 59…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078332 CAS číslo: 18190-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...