Tisk…

2-Ethynylpyridine 98% stabilizovaný

Dodavatel: Thermo Scientific
Varování


Stabilization: Stabilised with hydroquinone (HQ) 0,01%

L11274.03 L11274.06 L11274.14
L11274.03EA 1810 CZK
L11274.03 L11274.06 L11274.14
2-Ethynylpyridine 98% stabilizovaný
2-Ethynylpyridine

Stab. with 0.01% hydroquinone


Purity: 98% CAS No.: 1945-84-2 Molecular Formula: C7H5N Formula Weight: 103.12 Storage Temperature: 2°C to 8°C Flash Point: 76°(168°F) Refractive Index: 1.5590 Odour: Characteristic Boiling Point: 77-78°/14mm Beilstein Reference: 1098904 MDL No.: MFCD00041598

Vzorec: C₇H₅N
MW: 103,12 g/mol
Bod varu: 77…78 °C (14 mmHg)
Hustota: 1,020
Bod vzplanutí: 76 °C (168 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00041598
CAS číslo: 1945-84-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR