Tisk…

2-Bromotetradecane 95%

Dodavatel: Thermo Scientific

L10974.06 L10974.14
L10974.06EA 1200 CZK
L10974.06 L10974.14
2-Bromotetradecane 95%
2-Bromotetradecane

Purity: 95% CAS No.: 74036-95-6 Molecular Formula: C14H29Br Formula Weight: 277.30 Refractive Index: 1.4600 Boiling Point: 160-161°/8mm Beilstein Reference: 2071530 MDL No.: MFCD00045033

Vzorec: C₁₄H₂₉Br
MW: 277,30 g/mol
Bod varu: 160…161 °C (8 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00045033
CAS číslo: 74036-95-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR