Tisk…

2-Indanylacetic acid 99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-(2,3-Dihydro-1H-inden-2-yl)acetic acid

L10700.03 L10700.06
L10700.03EA 3300 CZK
L10700.03 L10700.06
2-Indanylacetic acid 99%
2-Indanylacetic acid

Purity: 99% CAS No.: 37868-26-1 Molecular Formula: C11H12O2 Formula Weight: 176.22 Melting Point: 91-93° Beilstein Reference: 2616409 MDL No.: MFCD00082653

Vzorec: C₁₁H₁₂O₂
MW: 176,22 g/mol
Bod tání: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00082653
CAS číslo: 37868-26-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR