1,2-O-Isopropylidene-ɑ-D-glucofuranose
Dodavatel: Thermo Scientific
Synonyms:
(R)-1-((3aR,5R,6S,6aR)-6-Hydroxy-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-5-yl)ethane-1,2-diol
L10688.06
L10688.14
L10688.06EA
985
CZK
L10688.06
L10688.14
1,2-O-Isopropylidene-ɑ-D-glucofuranose
1,2-O-Isopropylidene-alpha-D-glucofuranose
CAS No.: 18549-40-1
Molecular Formula: C9H16O6
Formula Weight: 220.22
Storage Temperature: 2°C to 8°C
Melting Point: 159-160°
Beilstein Reference: 82670
Optical Rotation: -13° (c=1 in water)
MDL No.: MFCD00063244
Vzorec:
C₉H₁₆O₆ MW: 220,22 g/mol Bod tání: 159…160 °C |
MDL Number:
MFCD00063244 CAS číslo: 18549-40-1 EINECS: 242-420-9 |