Tisk…

4,4'-Dimethoxychalcone 99%

Dodavatel: Thermo Scientific

Synonyms: 1,3-Bis(4-methoxyphenyl)prop-2-en-1-one

L10585.09 L10585.18
L10585.09EA 1890 CZK
L10585.09 L10585.18
4,4'-Dimethoxychalcone 99%
4,4'-Dimethoxychalcone

Purity: 99% CAS No.: 2373-89-9 Molecular Formula: C17H16O3 Formula Weight: 268.32 Melting Point: 99-103° Beilstein Reference: 1622293 MDL No.: MFCD00025815

Vzorec: C₁₇H₁₆O₃
MW: 268,32 g/mol
Bod tání: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00025815
CAS číslo: 2373-89-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR