Tisk…

Dimethyl adipimidate dihydrochloride 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Adipimidic Acid Dimethyl Ester Dihydrochloride

L10515.09 L10515.04
L10515.09EA 5190 CZK
L10515.09 L10515.04
Dimethyl adipimidate dihydrochloride 97%
Dimethyl adipimidate dihydrochloride

Purity: 97% CAS No.: 14620-72-5 Molecular Formula: C8H16N2O2.2HCl Formula Weight: 245.15 Melting Point: ca 215° dec. MDL No.: MFCD00012573

Vzorec: C₈H₁₆N₂O₂
MW: 245,14 g/mol
Bod tání: ∼ 215 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012573
CAS číslo: 14620-72-5
EINECS: 238-660-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR