1H,1H-Heptafluorobutyl acrylate 97% stabilizovaný
Dodavatel: Thermo Scientific
|
Varování
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 100 ppm
L10400.06
L10400.14
L10400.06EA
2710
CZK
L10400.06
L10400.14
1H,1H-Heptafluorobutyl acrylate 97% stabilizovaný
1H,1H-Heptafluorobutyl acrylate
Stab. with 100 ppm 4-methoxyphenol
Purity: 97%
CAS No.: 424-64-6
Molecular Formula: CH2=CHCO2CH2CF2CF2CF3
Formula Weight: 254.11
Flash Point: 31°(87°F)
Refractive Index: 1.3310
Odour: Pungent
Boiling Point: 57-58°/30mm
Beilstein Reference: 1792520
UN No.: UN3272
MDL No.: MFCD00039252
Vzorec:
C₇H₅F₇O₂ MW: 254,11 g/mol Bod varu: 57…58 °C (30 mmHg) Hustota: 1,418 Bod vzplanutí: 31 °C (87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039252 CAS číslo: 424-64-6 EINECS: 207-036-8 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...