Tetrahydrofurfuryl acrylate 90% stabilizovaný, tech.
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(Tetrahydrofuran-2-yl)methyl acrylate, Acrylic Acid Tetrahydrofurfuryl Ester
Stabilization:
Stabilised with hydroquinone (HQ) 500 ppm
L10299.18
L10299.30
L10299.18EA
1170
CZK
L10299.18
L10299.30
Tetrahydrofurfuryl acrylate 90% stabilizovaný, tech.
Tetrahydrofurfuryl acrylate
Stab. with 500ppm hydroquinone
Purity: 90%
CAS No.: 2399-48-6
Molecular Formula: C8H12O3
Formula Weight: 156.18
Flash Point: 93°(199°F)
Refractive Index: 1.4550
Boiling Point: 88-89°/9mm
Beilstein Reference: 113115
UN No.: UN3334
MDL No.: MFCD00014516
Vzorec:
C₈H₁₂O₃ MW: 156,18 g/mol Bod varu: 88…89 °C (9 mmHg) Hustota: 1,030 Bod vzplanutí: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014516 CAS číslo: 2399-48-6 EINECS: 219-268-7 UN: 3334 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...