Tisk…

Bis(2-dimethylaminoethyl)ether 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Dimethylaminoethyl Ether, 2,2'-Oxybis(N,N-dimethylethanamine)

L09884.09 L09884.18
L09884.09EA 2010 CZK
L09884.09 L09884.18
Bis(2-dimethylaminoethyl)ether 98%
Bis(2-dimethylaminoethyl)ether

Purity: 98% CAS No.: 3033-62-3 Molecular Formula: C8H20N2O Formula Weight: 160.26 Flash Point: 66°(151°F) Refractive Index: 1.4300 Boiling Point: 179-180° Beilstein Reference: 1739668 UN No.: UN2922 MDL No.: MFCD00059199

Vzorec: C₈H₂₀N₂O
MW: 160,26 g/mol
Bod varu: 179…180 °C
Hustota: 0,850
Bod vzplanutí: 66 °C (151 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059199
CAS číslo: 3033-62-3
EINECS: 221-220-5
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR