2-Tetrahydrofurfuryl isothiocyanate 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-(Isothiocyanatomethyl)tetrahydrofuran
L09878.06
L09878.03
L09878.06EA
4190
CZK
L09878.06
L09878.03
2-Tetrahydrofurfuryl isothiocyanate 98%
2-Tetrahydrofurfuryl isothiocyanate
Purity: 98%
CAS No.: 36810-87-4
Molecular Formula: C6H9NOS
Formula Weight: 143.21
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5350
Boiling Point: 122-125°/18mm
Beilstein Reference: 111696
UN No.: UN2922
MDL No.: MFCD00022476
Vzorec:
C₆H₉NOS MW: 143,21 g/mol Bod varu: 122…125 °C (18 mmHg) Hustota: 1,120 |
MDL Number:
MFCD00022476 CAS číslo: 36810-87-4 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...