DL-3-Phenyllactic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L09854.03
L09854.06
L09854.03EA
999
CZK
L09854.03
L09854.06
DL-3-Phenyllactic acid ≥98%
DL-3-Phenyllactic acid
Purity: 98+%
CAS No.: 828-01-3
Molecular Formula: C9H10O3
Formula Weight: 166.18
Odour: Odourless
Melting Point: 95-98°
Beilstein Reference: 2209791
MDL No.: MFCD00065928
Vzorec:
C₉H₁₀O₃ Bod tání: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065928 CAS číslo: 828-01-3 EINECS: 212-580-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...