2-Chloroethyl methacrylate 97% stabilised
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Stabilization:
400 - 600 ppm Stabilised with hydroquinone (HQ)
L09783.09
L09783.18
L09783.09EA
3510
CZK
L09783.09
L09783.18
2-Chloroethyl methacrylate 97% stabilised
2-Chloroethyl methacrylate
Stab. with 500ppm hydroquinone
Purity: 97%
CAS No.: 1888-94-4
Molecular Formula: C6H9ClO2
Formula Weight: 148.59
Flash Point: 69°(156°F)
Refractive Index: 1.4510
Boiling Point: 170°
Beilstein Reference: 956599
UN No.: UN2810
MDL No.: MFCD00045291
Vzorec:
C₆H₉ClO₂ MW: 148.586 g/mol Bod varu: 170 °C Bod vzplanutí: 69 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00045291 CAS číslo: 1888-94-4 EINECS: 217-566-1 UN: 2810 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...