Tisk…

1,4-Bis[(trimethylsilyl)ethynyl]benzene 98%

Dodavatel: Thermo Fisher Scientific

L09709.03 L09709.06
L09709.03EA 1100 CZK
L09709.03 L09709.06
1,4-Bis[(trimethylsilyl)ethynyl]benzene 98%
1,4-Bis[(trimethylsilyl)ethynyl]benzene

Purity: 98% CAS No.: 17938-13-5 Molecular Formula: C16H22Si2 Formula Weight: 270.53 Melting Point: 119-121° Beilstein Reference: 2840618 MDL No.: MFCD00135433

Vzorec: C₁₆H₂₂Si₂
MW: 270.53 g/mol
Bod tání: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00135433
CAS číslo: 17938-13-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR