Hexaketocyclohexane octahydrate 99%
Dodavatel: Thermo Fisher Scientific
L09488.09
L09488.04
L09488.09EA
7690
CZK
L09488.09
L09488.04
Hexaketocyclohexane octahydrate 99%
Hexaketocyclohexane octahydrate
Purity: 99%
CAS No.: 527-31-1
Molecular Formula: C6O6.8H2O
Formula Weight: 312.20 (168.07anhy)
Melting Point: 90-93°
Beilstein Reference: 2091040
MDL No.: MFCD00149074
Vzorec:
C₆O₆·8H₂O Bod tání: 90…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149074 CAS číslo: 7255-28-9 EINECS: 208-412-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...