Propargyl acrylate 96% stabilizovaný
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Prop-2-yn-1-yl acrylate
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 200 ppm
L09383.09
L09383.04
L09383.09EA
3160
CZK
L09383.09
L09383.04
Propargyl acrylate 96% stabilizovaný
Propargyl acrylate
Stab. with ca 200ppm BHT
Purity: 96%
CAS No.: 10477-47-1
Molecular Formula: C6H6O2
Formula Weight: 110.11
Storage Temperature: 2°C to 8°C
Flash Point: 43°(109°F)
Refractive Index: 1.4460
Odour: Pungent
Boiling Point: 138°
Beilstein Reference: 1902961
UN No.: UN3272
MDL No.: MFCD00078451
Vzorec:
C₆H₆O₂ MW: 110,11 g/mol Bod varu: 138 °C Hustota: 0,995 Bod vzplanutí: 43 °C (109 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00078451 CAS číslo: 10477-47-1 EINECS: 233-975-8 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...