4-Hydroxyphenyl benzoate 98%
Dodavatel: Thermo Scientific
|
Varování
|
L09350.06
L09350.14
L09350.06EA
1550
CZK
L09350.06
L09350.14
4-Hydroxyphenyl benzoate 98%
4-Hydroxyphenyl benzoate
Purity: 98%
CAS No.: 2444-19-1
Molecular Formula: C13H10O3
Formula Weight: 214.22
Melting Point: 164-167°
Beilstein Reference: 2050596
MDL No.: MFCD00053304
Vzorec:
C₁₃H₁₀O₃ MW: 214,22 g/mol Bod tání: 164…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053304 CAS číslo: 2444-19-1 EINECS: 219-479-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...