Tisk…

6-Methyl-3,5-heptadien-2-one 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 6-Methylhepta-3,5-dien-2-one

L09169.06 L09169.14
L09169.06EA 1200 CZK
L09169.06 L09169.14
6-Methyl-3,5-heptadien-2-one 97%
6-Methyl-3,5-heptadien-2-one

Purity: 97% CAS No.: 1604-28-0 Molecular Formula: C8H12O Formula Weight: 124.18 Refractive Index: 1.5340 Odour: Pleasant Boiling Point: 95-96°/20mm Beilstein Reference: 1699937 MDL No.: MFCD00043647

Vzorec: C₈H₁₂O
MW: 124.18 g/mol
Bod varu: 95…96 °C (20 mmHg)
Hustota: 0.890
Storage Temperature: Ambient
MDL Number: MFCD00043647
CAS číslo: 1604-28-0
EINECS: 216-507-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR