Tisk…

Ethyl-N,N-dimethyloxamate 98%

Dodavatel: Thermo Scientific

Synonyms: Ethyl 2-(dimethylamino)-2-oxoacetate

L08923.14 L08923.06
L08923.14EA 2170 CZK
L08923.14 L08923.06
Ethyl-N,N-dimethyloxamate 98%
Ethyl-N,N-dimethyloxamate

Purity: 98% CAS No.: 16703-52-9 Molecular Formula: C6H11NO3 Formula Weight: 145.16 Flash Point: 110°(230°F) Refractive Index: 1.4430 Boiling Point: 118-120°/10mm Beilstein Reference: 1906209 MDL No.: MFCD00015153

Vzorec: C₆H₁₁NO₃
MW: 145,16 g/mol
Bod varu: 118…120 °C (10 mmHg)
Hustota: 1,068
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015153
CAS číslo: 16703-52-9
EINECS: 240-755-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR