N-(tert-Butoxycarbonyl)-D-serine ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
N-Boc-D-serine, Boc-D-Ser-OH
L08904.03
L08904.06
L08904.03EA
1190
CZK
L08904.03
L08904.06
N-(tert-Butoxycarbonyl)-D-serine ≥98%
N-(tert-Butoxycarbonyl)-D-serine
Purity: 98+%
CAS No.: 6368-20-3
Molecular Formula: C8H15NO5
Formula Weight: 205.21
Storage Temperature: 2°C to 8°C
Melting Point: 88-91°
Beilstein Reference: 1874714
Optical Rotation: +3.5° (c=2 in acetic acid)
MDL No.: MFCD00063142
Vzorec:
C₈H₁₅NO₅ Bod tání: 88…91 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063142 CAS číslo: 6368-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...