2-Propylvaleryl chloride 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Propylpentanoyl chloride, 2,2-Di-n-propylacetylchloride
L08852.06
L08852.14
L08852.06EA
2310
CZK
L08852.06
L08852.14
2-Propylvaleryl chloride 98%
2-Propylvaleryl chloride
Purity: 98%
CAS No.: 2936-08-5
Molecular Formula: C8H15ClO
Formula Weight: 162.66
Boiling Point: 68-70°/10mm
Beilstein Reference: 1098856
UN No.: UN3265
MDL No.: MFCD00051446
Vzorec:
C₈H₁₅ClO MW: 162,66 g/mol Bod varu: 68…70 °C (10 mmHg) Hustota: 0,950 Storage Temperature: Ambient |
MDL Number:
MFCD00051446 CAS číslo: 2936-08-5 EINECS: 220-914-5 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...