2-(tert-Butoxycarbonyl)benzoic acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Phthalic acid mono-tert-butyl ester
L08534.06
L08534.14
L08534.06EA
3010
CZK
L08534.06
L08534.14
2-(tert-Butoxycarbonyl)benzoic acid 97%
2-(tert-Butoxycarbonyl)benzoic acid
Purity: 97%
CAS No.: 33693-84-4
Molecular Formula: C12H14O4
Formula Weight: 222.24
Melting Point: 84-88°
Beilstein Reference: 2263386
MDL No.: MFCD00043711
Vzorec:
C₁₂H₁₄O₄ MW: 222,24 g/mol Bod tání: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043711 CAS číslo: 33693-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...