5-Norbornene-2-carboxylic acid predominantly endo isomer 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid
L08444.06
L08444.14
L08444.06EA
821
CZK
L08444.06
L08444.14
5-Norbornene-2-carboxylic acid predominantly endo isomer 97%
5-Norbornene-2-carboxylic acid
Purity: 97%
CAS No.: 120-74-1
Molecular Formula: C8H10O2
Formula Weight: 138.17
Flash Point:
Vzorec:
C₈H₁₀O₂ MW: 138,17 g/mol Bod varu: 136…138 °C (14 mmHg) Hustota: 1,129 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00085356 CAS číslo: 120-74-1 EINECS: 204-422-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...