Tisk…

2-Chloro-5-methoxybenzimidazole 94%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Chloro-5-methoxy-1H-benzo[d]imidazole, 2-Chloro-5-methoxy-1H-benzimidazole

L08440.03 L08440.MD
L08440.03EA 6180 CZK
L08440.03 L08440.MD
2-Chloro-5-methoxybenzimidazole 94%
2-Chloro-5-methoxybenzimidazole

Purity: 94% CAS No.: 15965-54-5 Molecular Formula: C8H7ClN2O Formula Weight: 182.61 Melting Point: 165-168° Beilstein Reference: 777455 MDL No.: MFCD00051945

Vzorec: C₈H₇ClN₂O
MW: 182,607 g/mol
Bod tání: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00051945
CAS číslo: 15965-54-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR