N,N'-Diacetyl-1,4-phenylenediamine 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
DAPPD, N,N'-1,4-Phenylenebisacetamide, N,N'-(1,4-Phenylene)diacetamide
L08381.06
L08381.14
L08381.06EA
1810
CZK
L08381.06
L08381.14
N,N'-Diacetyl-1,4-phenylenediamine 98%
N,N'-Diacetyl-1,4-phenylenediamine
Purity: 98%
CAS No.: 140-50-1
Molecular Formula: C10H12N2O2
Formula Weight: 192.22
Melting Point:
Vzorec:
C₁₀H₁₂N₂O₂ Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026142 CAS číslo: 140-50-1 EINECS: 205-417-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...