Tisk…

Fluoral-P (=4-amino-3-penten-2-one) 96%

Dodavatel: Thermo Scientific

Synonyms: 4-Amino-3-penten-2-one, 4-Aminopent-3-en-2-one

L08379.06 L08379.03
L08379.06EA 2540 CZK
L08379.06 L08379.03
Fluoral-P (=4-amino-3-penten-2-one) 96%
Fluoral-P (=4-amino-3-penten-2-one)

Purity: 96% CAS No.: 1118-66-7 Molecular Formula: C5H9NO Formula Weight: 99.13 Melting Point: 36-41° Boiling Point: 209-213° Beilstein Reference: 1209344 MDL No.: MFCD00043715

Vzorec: C₅H₉NO
MW: 99,13 g/mol
Bod varu: 209…213 °C
Bod tání: 36…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00043715
CAS číslo: 1118-66-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR