Ethyl-2,4,6-trimethylphenylacetate 97%
Dodavatel: Thermo Scientific
Synonyms:
Ethyl mesitylacetate
L08275.09
L08275.04
L08275.09EA
6870
CZK
L08275.09
L08275.04
Ethyl-2,4,6-trimethylphenylacetate 97%
Ethyl-2,4,6-trimethylphenylacetate
Purity: 97%
CAS No.: 5460-08-2
Molecular Formula: C13H18O2
Formula Weight: 206.29
Refractive Index: 1.5050
Boiling Point: 135-140°/9mm
Beilstein Reference: 2105791
MDL No.: MFCD00051916
Vzorec:
C₁₃H₁₈O₂ MW: 206,285 g/mol Bod varu: 135…140 °C (9 mmHg) Hustota: 1,034 Storage Temperature: Ambient |
MDL Number:
MFCD00051916 CAS číslo: 5460-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...