Tisk…

Calcium-DL-glycerate hydrate

Dodavatel: Thermo Fisher Scientific

Synonyms: Calcium 2,3-Dihydroxypropionate, DL-Glyceric Acid Calcium Salt, 2,3-Dihydroxypropionic Acid Calcium Salt

L08214.03 L08214.06
L08214.03EA 2820 CZK
L08214.03 L08214.06
Calcium-DL-glycerate hydrate
Calcium-DL-glycerate hydrate

CAS No.: 67525-74-0 Molecular Formula: C6H10CaO8.2H2O Formula Weight: 286.26 (250.23anhy) Melting Point: ca 140° dec. Beilstein Reference: 4614104 MDL No.: MFCD00149289

Vzorec: C₃H₆O₄
MW: 250.22 g/mol
Bod tání: ∼ 140 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00149289
CAS číslo: 67525-74-0
EINECS: 282-247-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR