Tisk…

Methyl-β-D-galactopyranoside

Dodavatel: Thermo Fisher Scientific

Synonyms: Methyl-β-D-galactoside, (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol, (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

L08127.03 L08127.06
L08127.03EA 1530 CZK
L08127.03 L08127.06
Methyl-β-D-galactopyranoside
Methyl-beta-D-galactopyranoside

CAS No.: 1824-94-8 Molecular Formula: C7H14O6 Formula Weight: 194.18 Storage Temperature: 2°C to 8°C Melting Point: 175-179° Beilstein Reference: 81569 Optical Rotation: -16° (c=1.5 in methanol) MDL No.: MFCD00064357

Vzorec: C₇H₁₄O₆
Bod tání: 175…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064357
CAS číslo: 1824-94-8
EINECS: 217-361-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR