Methyl-2,5-dichlorothiophene-3-carboxylate 98%
Dodavatel: Thermo Scientific
|
Varování
|
L08118.03
L08118.06
L08118.03EA
2070
CZK
L08118.03
L08118.06
Methyl-2,5-dichlorothiophene-3-carboxylate 98%
Methyl-2,5-dichlorothiophene-3-carboxylate
Purity: 98%
CAS No.: 145129-54-0
Molecular Formula: C6H4Cl2O2S
Formula Weight: 211.07
Refractive Index: 1.5630
Boiling Point: 74-75°/0.6mm
MDL No.: MFCD00051763
Vzorec:
C₆H₄Cl₂O₂S MW: 211,07 g/mol Bod varu: 74…75 °C (0,6 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00051763 CAS číslo: 145129-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...