3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
3,3',5,5'-Tetra-tert-butyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione, 3,3',5,5'-Tetra-tert-butyldiphenoquinone
L07743.09
L07743.04
L07743.09EA
15270
CZK
L07743.09
L07743.04
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone ≥98%
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone
Purity: 98+%
CAS No.: 2455-14-3
Molecular Formula: C28H40O2
Formula Weight: 408.63
Melting Point: 242-246°
Beilstein Reference: 1916759
MDL No.: MFCD00051798
Vzorec:
C₂₈H₄₀O₂ MW: 408.63 g/mol Bod tání: 242…246 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051798 CAS číslo: 2455-14-3 EINECS: 219-527-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...