3-Butenoic acid 96%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Vinylacetic Acid, 3-Butenoic Acid
L07557.14
L07557.22
L07557.14EA
3610
CZK
L07557.14
L07557.22
3-Butenoic acid 96%
3-Butenoic acid
Purity: 96%
CAS No.: 625-38-7
Molecular Formula: C4H6O2
Formula Weight: 86.09
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.4240
Melting Point: -39°
Boiling Point: 162-163°
Beilstein Reference: 1699159
UN No.: UN3265
MDL No.: MFCD00002782
Vzorec:
C₄H₆O₂ Bod varu: 162…163 °C Bod tání: –39 °C Hustota: 1,012 Bod vzplanutí: 65 °C (149 °F) |
MDL Number:
MFCD00002782 CAS číslo: 625-38-7 EINECS: 210-892-5 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...