4-(Methylthio)-m-cresol 97%
Dodavatel: Thermo Fisher Scientific
L07353.06
L07353.14
L07353.06EA
783
CZK
L07353.06
L07353.14
4-(Methylthio)-m-cresol 97%
4-(Methylthio)-m-cresol
Purity: 97%
CAS No.: 3120-74-9
Molecular Formula: C8H10OS
Formula Weight: 154.23
Melting Point: 54-58°
Beilstein Reference: 1210322
MDL No.: MFCD00045773
Vzorec:
C₈H₁₀OS |
MDL Number:
MFCD00045773 CAS číslo: 3120-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...