Oxypurinol ≥98%
Dodavatel: Thermo Scientific
Synonyms:
4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 1H-Pyrazolo[3,4-d]pyrimidine-4,6-diol
L07160.03
L07160.06
L07160.MD
L07160.03EA
2920
CZK
L07160.03
L07160.06
L07160.MD
Oxypurinol ≥98%
Oxypurinol
Purity: 98+%
CAS No.: 2465-59-0
Molecular Formula: C5H4N4O2
Formula Weight: 152.11
Odour: Odourless
Melting Point:
Vzorec:
C₅H₄N₄O₂ MW: 152,11 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047205 CAS číslo: 2465-59-0 EINECS: 219-570-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...