Tisk…

Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate 98%

Dodavatel: Thermo Fisher Scientific

L06709.03 L06709.06
L06709.03EA 2460 CZK
L06709.03 L06709.06
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate 98%
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate

Purity: 98% CAS No.: 29113-33-5 Molecular Formula: C11H10N2O3 Formula Weight: 218.21 Melting Point: 189-193° Beilstein Reference: 190186 MDL No.: MFCD00051583

Vzorec: C₁₁H₁₀N₂O₃
Bod tání: 189…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00051583
CAS číslo: 29113-33-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR