Tisk…

2,2'-Dichlorobenzil ≥98%

Dodavatel: Thermo Scientific

Synonyms: 1,2-Bis(2-chlorophenyl)ethane-1,2-dione

L06507.06 L06507.14
L06507.06EA 2370 CZK
L06507.06 L06507.14
2,2'-Dichlorobenzil ≥98%
2,2'-Dichlorobenzil

Purity: 98+% CAS No.: 21854-95-5 Molecular Formula: C14H8Cl2O2 Formula Weight: 279.12 Melting Point: 132-136° Beilstein Reference: 2120689 MDL No.: MFCD00018263

Vzorec: C₁₄H₈Cl₂O₂
MW: 279,12 g/mol
Bod tání: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00018263
CAS číslo: 21854-95-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR