Glycolaldehyde diethyl acetal 98% stabilised
Dodavatel: Thermo Fisher Scientific
Varování
|
Synonyms:
Hydroxyacetaldehyde diethyl acetal, 2,2-Diethoxyethanol
Stabilization:
0.1% Stabilised with sodium hydroxide
Stab. with ca 0.1% sodium carbonate, 98%
Purity: 98%
CAS No.: 621-63-6
Molecular Formula: HOCH2CH(OCH2CH3)2
Formula Weight: 134.18
Storage Temperature: 2°C to 8°C
Flash Point: 62°(143°F)
Refractive Index: 1.4160
Boiling Point: 166-167°
Beilstein Reference: 773727
MDL No.: MFCD00051486
Vzorec:
HOCH₂CH(OCH₂CH₃)₂ MW: 134.18 g/mol Bod varu: 166…167 °C Bod vzplanutí: 62 °C (143 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00051486 CAS číslo: 621-63-6 EINECS: 210-697-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...