Tisk…

2-Butyn-1-al diethyl acetal 98%

Dodavatel: Thermo Fisher Scientific
Varování

L05872.03 L05872.06
L05872.03EA 1110 CZK
L05872.03 L05872.06
2-Butyn-1-al diethyl acetal 98%
2-Butyn-1-al diethyl acetal

Purity: 98% CAS No.: 2806-97-5 Molecular Formula: CH3C≡CCH(OCH2CH3)2 Formula Weight: 142.20 Flash Point: 56°(133°F) Refractive Index: 1.4260 Odour: Recognizable Boiling Point: 65-67°/12mm Beilstein Reference: 1702075 UN No.: UN1993 MDL No.: MFCD00009274

Vzorec: C₈H₁₄O₂
Bod varu: 65…67 °C (12 mmHg)
Hustota: 0,898
Bod vzplanutí: 56 °C (133 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009274
CAS číslo: 2806-97-5
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR