Ethyl-tert-butylacetate 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
tert-Butylacetic Acid Ethyl Ester, Ethyltert-butylacetate, 3,3-Dimethylbutyric Acid Ethyl Ester, Ethyl 3,3-Dimethylbutyrate
L05603.06
L05603.14
L05603.22
L05603.06EA
646
CZK
L05603.06
L05603.14
L05603.22
Ethyl-tert-butylacetate 98%
Ethyl-tert-butylacetate
Purity: 98%
CAS No.: 5340-78-3
Molecular Formula: (CH3)3CCH2CO2CH2CH3
Formula Weight: 144.22
Flash Point: 34°(93°F)
Refractive Index: 1.4035
Boiling Point: 144-146°
Beilstein Reference: 1749778
UN No.: UN3272
MDL No.: MFCD00026911
Vzorec:
C₈H₁₆O₂ MW: 144,21 g/mol Bod varu: 144…146 °C Hustota: 0,860 Bod vzplanutí: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026911 CAS číslo: 5340-78-3 EINECS: 226-280-6 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...