Tisk…

2-(4-Methoxybenzoyl)thiophene 97%

Dodavatel: Thermo Scientific

Synonyms: (4-Methoxyphenyl)(thiophen-2-yl)methanone, 4-Methoxyphenyl 2-Thienyl Ketone

L05513.09 L05513.18
L05513.09EA 2410 CZK
L05513.09 L05513.18
2-(4-Methoxybenzoyl)thiophene 97%
2-(4-Methoxybenzoyl)thiophene

Purity: 97% CAS No.: 4160-63-8 Molecular Formula: C12H10O2S Formula Weight: 218.27 Melting Point: 71-75° Beilstein Reference: 148560 MDL No.: MFCD00005440

Vzorec: C₁₂H₁₀O₂S
MW: 218,27 g/mol
Bod tání: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00005440
CAS číslo: 4160-63-8
EINECS: 223-997-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR