Tisk…

9-Aminofluorene hydrochloride ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 9-Fluorenamine Hydrochloride, 9H-Fluoren-9-amine hydrochloride

L05156.03 L05156.06
L05156.03EA 1040 CZK
L05156.03 L05156.06
9-Aminofluorene hydrochloride ≥98%
9-Aminofluorene hydrochloride

Purity: 98+% CAS No.: 5978-75-6 Molecular Formula: C13H11N.HCl Formula Weight: 217.70 Melting Point: ca 225° Beilstein Reference: 3915368 MDL No.: MFCD00012536

Vzorec: C₁₃H₁₁N
MW: 217,70 g/mol
Bod tání: ∼ 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00012536
CAS číslo: 5978-75-6
EINECS: 227-778-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR