Cinnamaldoxime mixture E/Z 98%
Dodavatel: Thermo Scientific
Synonyms:
Cinnamaldehyde oxime
L05026.06
L05026.14
L05026.06EA
1290
CZK
L05026.06
L05026.14
Cinnamaldoxime mixture E/Z 98%
Cinnamaldoxime
Purity: 98%
CAS No.: 13372-81-1
Molecular Formula: C9H9NO
Formula Weight: 147.18
Melting Point: 117-122°
Beilstein Reference: 3117555
MDL No.: MFCD00019969
Vzorec:
C₉H₉NO MW: 147,18 g/mol Bod tání: 117…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019969 CAS číslo: 13372-81-1 EINECS: 236-449-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...