Tisk…

N-Methylpentylamine 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: N-Pentylmethylamine

L04882.04 L04882.09
L04882.04EA 3300 CZK
L04882.04 L04882.09
N-Methylpentylamine 98%
N-Methylpentylamine

Purity: 98% CAS No.: 25419-06-1 Molecular Formula: CH3(CH2)4NHCH3 Formula Weight: 101.19 Flash Point: 18°(64°F) Refractive Index: 1.4105 Boiling Point: 117-119° Beilstein Reference: 1731333 UN No.: UN2733 MDL No.: MFCD00041354

Vzorec: C₆H₁₅N
MW: 101,19 g/mol
Bod varu: 117…119 °C
Hustota: 0,738
Bod vzplanutí: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041354
CAS číslo: 25419-06-1
EINECS: 246-966-9
UN: 2733
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR