Tisk…

Trimethylsilylmethanol 95%

Dodavatel: Thermo Fisher Scientific
Varování

L04251.03 L04251.06
L04251.03EA 1260 CZK
L04251.03 L04251.06
Trimethylsilylmethanol 95%
Trimethylsilylmethanol

Purity: 98+% CAS No.: 3219-63-4 Molecular Formula: (CH3)3SiCH2OH Formula Weight: 104.23 Flash Point: 31°(87°F) Refractive Index: 1.4190 Boiling Point: 121-122° Beilstein Reference: 1731486 UN No.: UN1993 MDL No.: MFCD00004596

Vzorec: C₄H₁₂OSi
Bod varu: 121…122 °C
Hustota: 0,827
Bod vzplanutí: 31 °C (87 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004596
CAS číslo: 3219-63-4
EINECS: 221-746-5
ADR: 3,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR