Tisk…

4-(2-Chloroethyl)benzoic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

L04068.09 L04068.18
L04068.09EA 3330 CZK
L04068.09 L04068.18
4-(2-Chloroethyl)benzoic acid 97%
4-(2-Chloroethyl)benzoic acid

Purity: 97% CAS No.: 20849-78-9 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 187-189° Beilstein Reference: 2084041 MDL No.: MFCD00013995

Vzorec: C₉H₉ClO₂
MW: 184.62 g/mol
Bod tání: 187…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00013995
CAS číslo: 20849-78-9
EINECS: 244-080-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR