Vinyl(chloromethyl)dimethylsilane 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L04056.14
L04056.06
L04056.14EA
7010
CZK
L04056.14
L04056.06
Vinyl(chloromethyl)dimethylsilane 97%
Vinyl(chloromethyl)dimethylsilane
Purity: 97%
CAS No.: 16709-86-7
Molecular Formula: C5H11ClSi
Formula Weight: 134.68
Flash Point: 14°(57°F)
Refractive Index: 1.4430
Boiling Point: 121-122°
Beilstein Reference: 2071374
UN No.: UN1993
MDL No.: MFCD00040860
Vzorec:
C₅H₁₁ClSi MW: 134.68 g/mol Bod varu: 121…122 °C Hustota: 0.893 Bod vzplanutí: 14 °C (57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040860 CAS číslo: 16709-86-7 EINECS: 240-758-1 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...